Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥90.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Acrylic Acid Hydroxypropyl Ester, Propylene Glycol Monoacrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A0744-25ML A0744-500ML
TCIAA0744-25MLEA 132 DKK
TCIAA0744-25ML TCIAA0744-500ML
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate) ≥90.0% (by GC) stabilised
Hydroxypropyl acrylate (mixture of 2-hydroxypropyl and 2-hydroxy-1-methylethyl acrylate)
Formel: C₆H₁₀O₃
Flammepunkt: 100°C
MDL Number: MFCD04113589
CAS-nummer: 25584-83-2
EINECS: 247-118-0
UN nummer: 2922
ADR: 8,II

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 % (total of isomers )


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR