Acid blue 119 (Alkali blue 6B)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Thionine blue, [[4-[Hydroxybis[4-(phenylamino)phenyl]methyl]phenyl]amino]benzenesulphonic acid monosodium salt, Alkali Blue 6B

A0596-1G A0596-5G
TCIAA0596-1GEA 238 DKK
TCIAA0596-1G TCIAA0596-5G
Acid blue 119 (Alkali blue 6B)
Acid blue 119 (Alkali blue 6B)
Formel: C₃₇H₃₀N₃NaO₄S
MDL Number: MFCD00083221
CAS-nummer: 1324-80-7

Bestil nu

Specifikationer Testresultater

Appearance Blue to Brown to Dark blue powder to crystal
Lambda max. 599.0 to 604.0 nm (MeOH)
Absorbance min. 0.400 (5mg/L, MeOH, 599.0 to 604.0 nm) (calcd.on dried substance)
Drying loss max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR