Agmatine sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Amino-4-guanidobutane Sulphate, 1-(4-Aminobutyl)guanidine sulphate

A0310-1G A0310-100MG
TCIAA0310-1GEA 542 DKK
TCIAA0310-1G TCIAA0310-100MG
Agmatine sulphate ≥98.0% (by titrimetric analysis)
Agmatine sulphate
Formel: C₅H₁₄N₄
Molekylvægt: 228.27 g/mol
MDL Number: MFCD00013109
CAS-nummer: 2482-00-0
EINECS: 219-617-3
Merck Index: 14,00188

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR