Bis(aminoguanidinium)sulfat ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Bis(1-aminoguanidinium) sulphate, Aminoguanidine hemisulphate salt, Aminoguanidine sulphate hydrate

A0309-25G A0309-500G
TCIAA0309-25GEA 173 DKK
TCIAA0309-25G TCIAA0309-500G
Bis(aminoguanidinium)sulfat ≥98.0% (by titrimetric analysis)
Bis(aminoguanidinium)sulfat
Formel: C₂H₁₄N₈O₄S
Molekylvægt: 246.25 g/mol
MDL Number: MFCD00035665
CAS-nummer: 996-19-0
EINECS: 213-628-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9bedd43f0b280c44ed2260e84f2be069ad3ae9be&vid=b9b46065560d46a19da206426834f965a18a5acc&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Potassium iodate Method) min. 98.0% (calcd.on anh.substance)
Solubility in Water almost transparency
Water max. 8.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR