4-Amino-4'-(N,N-dimethylamino)stilbene ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N,N-Dimethyl-4,4'-stilbenediamine, 4-(4-Aminostyryl)-N,N-dimethylaniline

A0291-5G A0291-1G
TCIAA0291-5GEA 3350 DKK
TCIAA0291-5G TCIAA0291-1G
4-Amino-4'-(N,N-dimethylamino)stilbene ≥98.0% (by titrimetric analysis)
4-Amino-4'-(N,N-dimethylamino)stilbene
Formel: C₁₆H₁₈N₂
Molekylvægt: 238.33 g/mol
MDL Number: MFCD00059189
CAS-nummer: 22525-43-5

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 170.0 to 174.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR