1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Amino-4-bromo-9,10-dihydro-9,10-dioxo-2-anthracenesulfonic acid sodium salt, 1-Amino-4-bromoanthraquinone-2-sulphonic Acid Sodium Salt, Bromaminic acid Sodium Salt, Bromaminic acid sodium salt, Sodium 1-amino-4-bromo-9,10-dioxo-9,10-dihydroanthracene-2-sulphonate
A0279-25G
A0279-500G
TCIAA0279-25GEA
132
DKK
TCIAA0279-25G
TCIAA0279-500G
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt ≥98.0% (by HPLC)
1-Amino-4-bromoanthraquinone-2-sulphonic acid sodium salt
Formel:
C₁₄H₈BrNO₅S Molekylvægt: 404.17 g/mol |
MDL Number:
MFCD00019160 CAS-nummer: 6258-06-6 EINECS: 228-391-5 |
Specifikationer Testresultater
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 93.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...