Stainless Steel Buckets, Graduated
Leverandør: Sampling Systems
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
A264-15 A264-10 A264-10L A264-12 A264-12L A264-15L
SAMPA264-15EA 1790 DKK
SAMPA264-15 300-0649 300-0650 300-0651 300-0652 300-0653
Stainless Steel Buckets, Graduated
Spande
Stainless steel, graduated buckets are ideal for use in food, chemical, and general industry.
  • 304 stainless steel construction
  • Skirtband gives extra protection to the base
  • Robust design
  • Easy to use
Graduated stainless steel buckets are made from heavy gauge stainless steel and are very durable. They are ideal for general-purpose factory work. The swing handle allows them to be lifted easily. Optional lids are available separately.
The 10,12 and 15 litre buckets feature a skirt band around the base. This provides extra protection to the base and increases stability.