1-Cetylpyridiniumchlorid monohydrat 96.0-101.0%
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Hexadecylpyridinium chloride monohydrate, 1-Hexadecylpyridinium chloride monohydrate, Cetylpyridinium Chloride, Monohydrate, Cetylpyridinium chloride monohydrate
226991000 226995000 226990025
ACRO226991000EA 454 DKK
ACRO226991000 ACRO226995000 ACRO226990025
1-Cetylpyridiniumchlorid monohydrat 96.0-101.0%
1-Cetylpyridiniumchlorid monohydrat
Formel: C₂₁H₃₈ClN·1H₂O
Molekylvægt: 358.01 g/mol
Smeltepunkt: 80…83 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149977
CAS-nummer: 6004-24-6
EINECS: 204-593-9
UN nummer: 2811
ADR: 6.1 II
Merck Index: 13,02041
Bestil nu
Specifikationer Testresultater
Appearance White Powder
Infrared spectrum Conforms
Water 4.5 to 5.5 %
Titration 96.0 to 101.0 %