Jern(III)sulfat-hydrat, Reagent Grade

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

33316.A7 33316.30 33316.A1
33316.A7EA 4940 DKK
33316.A7 33316.30 33316.A1
Jern(III)sulfat-hydrat, Reagent Grade
Jern(III)sulfat-hydrat

CAS No.: 15244-10-7 Molecular Formula: Fe2(SO4)3.xH2O Formula Weight: 399.87(anhy) Physical Form: Granular/Powder Density: 2.0-2.1 Sensitivity: Hygroscopic Appearance: Grayish white Solubility: Soluble in water MDL No.: MFCD00011007

Formel: Fe₂(SO₄)₃·xH₂O
Massefylde: 2,0…2.1
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149714
CAS-nummer: 15244-10-7
EINECS: 233-072-9
Merck Index: 13,04065

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR