(S)-(-)-Aminopyrrolidin ≥99%, ee 99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

L19500.03 L19500.06
L19500.03EA 1280 DKK
L19500.03 L19500.06
(S)-(-)-Aminopyrrolidin ≥99%, ee 99%
(S)-(-)-Aminopyrrolidin

Purity: 99% CAS No.: 128345-57-3 Molecular Formula: C4H10N2 Formula Weight: 86.14 Flash Point: 68°(154°F) Refractive Index: 1.4880 Boiling Point: 159-160° Beilstein Reference: 4290648 Optical Rotation: -20° (neat) UN No.: 2735 MDL No.: MFCD00143193

Formel: C₄H₁₀N₂
Molekylvægt: 86.14 g/mol
Kogepunkt: 159…160 °C (1013 hPa)
Massefylde: 0.98 g/cm³ (20 °C)
Flammepunkt: 68 °C (154 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00143193
CAS-nummer: 128345-57-3

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR