(R)-(+)-Aminopyrrolidin ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (R)-Pyrrolidin-3-amine

A1167.0025 A1167-5G A1167-1G
TCIAA1167.0025EA 0 DKK
TCIAA1167.0025 TCIAA1167-5G TCIAA1167-1G
(R)-(+)-Aminopyrrolidin ≥98.0% (by GC, titration analysis)
(R)-(+)-Aminopyrrolidin
Formel: C₄H₁₀N₂
Molekylvægt: 86.14 g/mol
Flammepunkt: 64°C
MDL Number: MFCD00143190
CAS-nummer: 116183-82-5
UN nummer: 2735
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=555dc9d053cf176f12cfae06a11c171717b37833&vid=555dc9d053cf176f12cfae06a11c171717b37833&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 99.0 ee%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR