Tetraethylammoniumfluorid, hydrat ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

B23173.06 B23173.14 B23173.22
B23173.06EA 813 DKK
B23173.06 B23173.14 B23173.22
Tetraethylammoniumfluorid, hydrat ≥97%
Tetraethylammoniumfluorid, hydrat

Purity: 97% CAS No.: 98330-04-2 Molecular Formula: (CH3CH2)4NF.xH2O Formula Weight: 149.26(anhy) Beilstein Reference: 4152659 MDL No.: MFCD00149990

Formel: C₈H₂₀FN·xH₂O
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149990
CAS-nummer: 98330-04-2
EINECS: 211-559-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR