L-Lysin

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

28195.01 28195.02 28195.03
SERA28195.01EA 290.95 DKK
SERA28195.01 SERA28195.02 SERA28195.03
L-Lysin
L-Lysin
Formel: C₆H₁₄N₂O₂
Kogepunkt: 290 °C (1013 hPa)
Smeltepunkt: 196 °C
MDL Number: MFCD00064433
CAS-nummer: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR