L(+)-Serin for synthesis, Sigma-Aldrich®

Leverandør: Merck
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

M-Clarity™ quality level = MQ200

8.16020.0025 8.16020.0100
8.16020.0025EA 369 DKK
8.16020.0025 8.16020.0100
L(+)-Serin for synthesis, Sigma-Aldrich®
L(+)-Serin
Formel: C₃H₇NO₃
Molekylvægt: 105.09 g/mol
Kogepunkt: 259
Smeltepunkt: 215…225 °C
Massefylde: 1.6 g/cm³ (22 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064224
CAS-nummer: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Bestil nu

Specifikationer Testresultater

Solubility 364 g/l (20 °C)
Melting Point 215 - 225 °C decomposes
Molar Mass 105.09 g/mol
Bulk Density 570 kg/m3
pH 5 - 6 (100 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR