MES monohydrat
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
MES monohydrate, 2-(N-Morpholino)ethanesulphonic acid monohydrate, 2-Morpholinoethanesulphonic acid hydrate
29830.01
29830.02
29830.03
SERA29830.01EA
0
DKK
SERA29830.01
SERA29830.02
SERA29830.03
MES monohydrat
MES monohydrat
Formel:
C₆H₁₃NO₄S·1H₂O Molekylvægt: 213.25 g/mol Smeltepunkt: > 300 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149409 CAS-nummer: 145224-94-8 EINECS: 224-632-3 Merck Index: 13,05929 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...