Zink 1.000 mg/l i fortyndet salpetersyre (from Zn) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456384KEA
2220
DKK
456384K
456382Y
Zink 1.000 mg/l i fortyndet salpetersyre (from Zn) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Zinc Standards
Zn in HNO₃ 2 to 5%.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.