Cobaltstandard, 1g/l Co i salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86670.260EA 230.3 DKK
86670.260 86670.180
Cobaltstandard, 1g/l Co i salpetersyre AVS TITRINORM standard til AAS
Standarder Kationiske standarder Cobalt Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%