Kalium 1.000 mg/l i fortyndet salpetersyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86686.260EA 235.2 DKK
86686.260 86686.180
Kalium 1.000 mg/l i fortyndet salpetersyre AVS TITRINORM standard til AAS
Standarder Kationiske standarder Potassium Standards
  • Traceable to SRM from NIST
  • Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
  • Supplied with certificate of analysis, real value ±0,3%