Iridium 1.000 mg/l i 10% saltsyre (from IrCl3) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455502REA
2690
DKK
455502R
455504T
Iridium 1.000 mg/l i 10% saltsyre (from IrCl3) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Iridium Standards
IrCl₃.3H₂O in HCl 10%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.