Holmium 10.000 mg/l i fortyndet salpetersyre (de Ho2O3) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455472FEA
4430
DKK
455472F
455474H
Holmium 10.000 mg/l i fortyndet salpetersyre (de Ho2O3) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Holmium Standards
Ho₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.