Beryllium 1.000 mg/l i fortyndet salpetersyre, ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455082SEA
1330
DKK
455082S
455084U
Beryllium 1.000 mg/l i fortyndet salpetersyre, ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Beryllium Standards
BeO.(Be(OAc)₂)₃) in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.