Saltsyre ≥33% ≥99.999999% (metals basis)
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Purity: 99.999999%
CAS No.: 7647-01-0
Molecular Formula: HCl
Formula Weight: 36.46
Physical Form: Liquid, double distilled in quartz
Density: 1.19
Appearance: Colourless to pale yellow
Odour: Pungent
Boiling Point: 81.5-110°
Merck Reference: 14,4780
UN No.: 1789
MDL No.: MFCD00011324
Formel:
HCl Kogepunkt: 81,5…110 °C Smeltepunkt: –30 °C Massefylde: 1,19 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011324 CAS-nummer: 7647-01-0 EINECS: 231-595-7 UN nummer: 1789 ADR: 8,II Merck Index: 14,4780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...