Ethanol-D6 (99% D)
Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Methyl carbinol-D6, Hydroxy ethane-D6, Ethanol-[²H₆], Ethyl alcohol-D6
DLM-31-5X1
DLM-31-BULK
CAMBDLM-31-5X1EA
0
DKK
CAMBDLM-31-5X1
CAMBDLM-31-BULK
Ethanol-D6 (99% D)
Ethanol-D6
Formel:
D₃CCD₂OD Molekylvægt: 52.02 g/mol Kogepunkt: 78 Smeltepunkt: –114.5 °C Massefylde: 0.801 g/cm³ (20 °C) Flammepunkt: 13 |
MDL Number:
MFCD00051020 CAS-nummer: 1516-08-1 EINECS: 216-162-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...