Petroleumbenzin, 100-140 °C, Puriss.

Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Gasoline, Petroleum ether, Petrol, Benzine

24534-1L 24534-2.5L 24534-25L 24534-4X2.5L
24534-1LEA 538 DKK
24534-1L 24534-2.5L 24534-25L 24534-4X2.5L
Petroleumbenzin, 100-140 °C, Puriss.
Petroleumbenzin

RTECS Number: OI6180000
UNSPSC Code: 12191502

Kogepunkt: 90…165 °C (1,013 hPa)
Smeltepunkt: –40 °C
Massefylde: 0,725…0.748 g/cm³ (20 °C)
Flammepunkt: <10 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081849
CAS-nummer: 64742-49-0
EINECS: 920-750-0
UN nummer: 3295
ADR: 3,II

Bestil nu

Specifikationer Testresultater

Boiling range 100 - 140 °C
Density (D 20/4) 0.720 - 0.745
Refractive index (n 20/D) 1.403 - 1.412
Non-volatile matter Max. 0.005 %
Water (Karl Fischer) Max. 0.01 %
Aromatic substances (as C₆H₅CH₃) Max. 0.02 %
S-compounds (as S) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR