Let destillat suitable for use in the testing of petroleum products by IP and ASTM methods
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Destillater (råolie), hydrogenbehandlede lette
286735QEA 988 DKK
286735Q 28673LJ
Let destillat suitable for use in the testing of petroleum products by IP and ASTM methods
Let destillat
Kogepunkt: 192…256 °C (760 mmHg Hg)
Massefylde: 0,805 g/cm³ (15 °C)
Flammepunkt: 77 °C (closed cup)
CAS-nummer: 64742-47-8
EINECS: 265-149-8
REACH: 01-2119484819-18
Bestil nu
Specifikationer Testresultater
Aromatics (W/V) Max. 1.00 %
Distillation range (98 %) 185 - 280°C