Counting chambers, Fuchs-Rosenthal
Leverandør: GLASWARENFABRIK KARL HECHT
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40449702 40449703
631-0929EA 443 DKK
631-0929 631-0924
Counting chambers, Fuchs-Rosenthal
Objektglas Tællekamre
This ruling is used for counting leukocytes, eosinophils and cells in cerebro-spinal fluid.
  • Total area of ruling: 16 mm²
  • Subdivision: 16 square groups of 1 mm² each
  • 16 mini squares (0,25×0,25 mm) each