VWR® Advanced LPG / LPCG / LPWG, Precision Balances with Graphic Display
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
611-2306EA 6400 DKK
611-2306 611-2291 611-2297 611-2310 611-2307 611-2285 611-2287 611-2283 611-2373 611-2372 611-3273 611-3274 611-3275 611-3276 611-3278 611-3284 611-3358 611-3365 611-3366 611-3381 611-3390 611-3393 611-3460 611-3474 611-3832 611-3833 611-4318 611-4329 611-4344 611-4345 611-4346 611-4347 611-4348 611-5048
VWR® Advanced LPG / LPCG / LPWG, Precision Balances with Graphic Display
Vægte
Precision balances that are ideal for use in any laboratory, these models offer GLP/ISO records of weight values with date, time and serial number. A series that ranges from simple weighing to operation with automated internal calibration. All Advanced models have a graphic display with adjustable contrast, an ergonomic keypad and a stainless steel top pan.
  • Selectable filter levels and user can select the measuring unit that the balance will use to display the weight
  • RS232 data interface
  • GLP set-up - stores balance, project and user identification data, as well as date and time
  • Easy to operate and in multiple languages: English, German, French, Italian, Spanish and Portuguese
Functions include suspended weighing, piece counting, percentage weighing, check weighing, density determination, totalisation, formulation and maximum load (peak hold) functions.
Users can choose three options for models with internal calibration (denoted by 'i' at end of the model name):
Balance will calibrate using the internal weight by pressing CAL button.
Balance will automatically recalibrate when ambient temperature changes and/or after a certain time has passed (Incal+).
Users can choose to calibrate the balance with an external weight.
Statistic printouts are available if connected to a statistical printer. Under balance weighing hooks are available for several models. The LPWG models have a graphic display and three door draft shield, LPCG models have a graphic display and circular draft shield.
Certificeringer: EN 61326-1:2013, EN 61010-1: 2010, EN 61010-2-010:2014, EN 50581:2012. Comply with the European Directive 2014- 31-EU (NAWI) and Harmonised Standard EN 45501 (verified, approved).
Bestillingsoplysninger: Balances with suffix 'D' have dual-range, suffix i denotes internal calibration. Models with suffix 'C' are delivered with factory calibration certificate, suffix 'M' denotes approved or verified models and suffix 'P' denotes units are complete with trap kit for pipette calibration. Models with prefix 'C' have a circular draft shield and prefix 'W' indicates a three-door draft shield.
Leveringsinformation: Supplied with a plastic in use protective cover. A wide range of accessories including printer, software and PC connection cable are available and must be purchased separately.