VWR®, Diesel and lubricant, multi-parameter Certified Reference Materials (MPCRM)
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
99101.260EA 4000 DKK
99101.260 99100.260
VWR®, Diesel and lubricant, multi-parameter Certified Reference Materials (MPCRM)
Standarder Petroleumsindustriens standarder
MPCRM’s provide both calibration and verification options for laboratory test equipment, all from the same sample, with their values certified to the highest level of accreditation integrity available globally.
  • Highest level of accreditation guarantee, providing the most credible certified data currently available worldwide
  • Low levels of uncertainty achieves maximum accuracy of data
  • Multiple test parameters provide a flexible and cost effective solution
  • Fully traceable to international standards
  • Glass packaging with tamper-evident cap
Diesel: Certified for Viscosity, Density, Flash Point, Cloud Point, CFPP and Distillation using ASTM / IP and EN test method protocols
Lubricant: Certified for Viscosity, Density, Flash Point and Pour Point using ASTM / IP and EN test method protocols
Certificeringer: Fully dual-certified in accordance with ISO 17025 and ISO 17034 under accreditation by UKAS.
Advarsel: The ordering table below provides a small sample of the data availble with documentation.