CF® Dye dCTP conjugates
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40028 40027 40058 40055 40056 40057 40066 40067 40068 40027-T 40028-T 40055-T 40056-T 40057-T 40058-T 40066-T 40067-T 40068-T
40028.EA 4620 DKK
40028. 40027. 40058. 40055. 40056. 40057. 40066. 40067. 40068. 40027-T 40028-T 40055-T 40056-T 40057-T 40058-T 40066-T 40067-T 40068-T
CF® Dye dCTP conjugates
Proteinkoblings- og mærkningssystemer
CF® dye-dCTP can be used to synthesise probes for in situ hybridisation, microarray, or to synthesise labelled DNA probes for in situ hybridisation and nucleic acid blotting applications.
  • Synthesise exceptionally bright and photostable fluorescent DNA probes
  • Detect by fluorescence microscopy, flow cytometry, or in gel
  • Compatible with fixed cells or tissue sections
  • Choice of nine CF® dye colours from green to far-red
  • CF® dyes have superior brightness, photostability, and are water-soluble
Advarsel: CF™594 dCTP is not compatible with PCR labelling of DNA probes.