Thermo Scientific™ Nalgene™ PC certified clean biotainer carboys
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
383030-42 383120-42 383233-42
216-2278EA 9010 DKK
216-2278 216-2279 216-2281
Thermo Scientific™ Nalgene™ PC certified clean biotainer carboys
Flasker Flasker, dunke
The Nalgene™ multi-tiered portfolio provides bioproduction facilities with a variety of high-quality, sterile storage and transport solutions. Choose the appropriate product necessary to suit any application, through the step-down process and into the cleanroom.
  • Sterile
  • Clear finish
  • Impact resistant polycarbonate construction
These carboys can reduce the risk of contamination.
Closure size: 48 mm
Certificeringer: Validated to USP 788 and <0,5 EU/ml endotoxin.