Tyramide amplification kit with HRP streptavidin and CF® dye or biotin tyramide
Leverandør: Biotium
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33002 33005 33008 33011 33014 33017 33020
33002.EA 6280 DKK
33002. 33005. 33008. 33011. 33014. 33017. 33020.
Tyramide amplification kit with HRP streptavidin and CF® dye or biotin tyramide
Proteinkoblings- og mærkningssystemer
Kits for increasing immunofluorescence sensitivity using HRP-catalysed tyramide signal amplification with the choice of next-generation fluorescent CF® dyes or biotin.
  • Increase sensitivity by up to 100-fold
  • Use lower primary antibody concentrations
  • Boost signal in challenging samples, like human FFPE tissues
  • Choose from six bright and photostable CF® dyes or biotin
Tyramide amplification, sometimes called Catalysed Reporter Deposition (CARD), is an enzyme-mediated detection method that uses the catalytic activity of horseradish peroxidase (HRP) to generate high density labelling of a target protein or nucleic acid sequence in situ. The tyramide amplification method has been reported to increase the sensitivity by up to 100-fold compared to conventional avidin-biotin detection. Multiple rounds of tyramide signal amplification can be performed for multiplex detection.
These kits includes choice of six CF® dye tyramides or biotin tyramide. CF® dyes are line of next-generation fluorescent dyes, with advantages in brightness and photostability compared to other dyes.
Bestillingsoplysninger: Each kit contains reagents for staining 50 to 150 slides; the actual number of samples that can be stained will depend on staining volumes used. Tyramide amplification buffer, CF® dye tyramides, and other tyramide compounds are also available separately. Note: Endogenous biotin blocking reagents (not provided) may be required for streptavidin/biotin detection in cells or tissues.
Leveringsinformation: Kit components: CF® dye or biotin-tyramide stock solution, HRP Streptavidin, BSA for blocking, Tyramide amplification buffer plus, 30% Hydrogen peroxide.