SafeKey compact lockout padlocks, nylon
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
150183 150184 150185 150186 150187 150188 150189 150190 150191 150192 150193 150194 150195 150196 150197 150207 150208 150209 150210 150211 150212 150213 150214 150215 150180 150181 150182
BRDY150183EA 133 DKK
BRDY150183 BRDY150184 BRDY150185 BRDY150186 BRDY150187 BRDY150188 BRDY150189 BRDY150190 BRDY150191 BRDY150192 BRDY150193 BRDY150194 BRDY150195 BRDY150196 BRDY150197 BRDY150207 BRDY150208 BRDY150209 BRDY150210 BRDY150211 BRDY150212 BRDY150213 BRDY150214 BRDY150215 BRDY150180 BRDY150181 BRDY150182
SafeKey compact lockout padlocks, nylon
Sikkerhedslåse
These SafeKey nylon compact padlocks are a small and lightweight range of padlocks suitable for use in energy isolation programs or in small spaces.
  • Lightweight with a ribbed design for easy gripping
  • Ideal for electrical applications and enclosures
  • Key releases only upon lock closure
  • Preferred for most indoor lockout applications
  • Engraving options on front, back, left side, right side, and/or key
  • Non-conductive
The new design and innovative keying mechanism provides more unique key possibilities than a conventional standard safety padlock, without sacrificing a secure lockout. The unique lock mechanism is insulated and non-conductive to protect workers from shocks when key is inserted. The lock mechanism has six precision steel tumbler blades for a high number of key differences and tamper resistance.
Locks are compact and lightweight and resist impact, temperature, chemicals, and corrosion. Their non-conductive, non-sparking body features a ribbed design for easier gripping and handling. The colour-matched key and lock body allows a fast identification in the application.
Bestillingsoplysninger: Custom keying, engraving, charting, Master Key, and Grand Master Key options are available.
Leveringsinformation: One key is supplied with each lock.