Box sealing tapes 3739, Scotch®
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3739B756 3739B 3739C
817-0444EA 1690 DKK
817-0444 817-0443 817-0446
Box sealing tapes 3739, Scotch®
Tape
These tapes feature a high strength polypropylene film backing and hot melt synthetic rubber resin adhesive.
  • Re-seal and package heavy weights of valuable goods
  • High initial tack and shear adhesion to fibreboard
  • Use for single strip closure of fibreboard boxes
  • Performs well at all temperatures typically encountered by packaged products in shipping and storage environments
  • Tape resists splitting when subjected to rough handling and absorbs impact shock
  • Tensile strength: 135 [Newton/25 mm]
  • BOPP back material
Mainly used for single-strip closure of fibreboard boxes, re-sealing, and packaging of heavyweight or valuable goods. This tape performs well throughout all temperatures typically encountered by packaged products in shipping and storage environments.