Adhesive transfer tapes, ATG 969, Scotch®
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
969-12MM 969-19MM
MMMA969-12MMEA 20360 DKK
MMMA969-12MM MMMA969-19MM
Adhesive transfer tapes, ATG 969, Scotch®
Tape
Use Scotch® ATG adhesive transfer tape 969 as a safe alternative to hot melt adhesives. This tape features high initial adhesion and good holding power to a variety of materials including most plastics. It is an acrylic, reverse wound, pressure sensitive adhesive on a densified kraft paper liner that can be used with Scotch® ATG adhesive applicators.
  • Bonds well to smooth and moderately-textured surfaces
  • Cost effective replacement to rivets, staples, screws, bolts, or liquid adhesives
  • Conforms easily to curves and contours
  • Leaves no mess, no residue and no odour
  • Length: 33 m
  • Transfer tape back material
Get an acrylic, reverse wound, pressure sensitive adhesive on a densified kraft paper liner with Scotch® ATG adhesive transfer tape 969. The ATG tape system delivers a controlled application of the adhesive while the liner rewinds into the ATG applicator. It bonds on contact, and works on a variety of substrate or material combinations. The ATG tape system is versatile, convenient, quick, and easy to use. There is no mess or clean up. It is a cost effective replacement to rivets, staples, screws, bolts, or liquid adhesives.