Adhesive transfer tapes, ATG 924, Scotch®
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
924EU-12MM 924EU-19MM
MMMA924EU-12MMEA 7510 DKK
MMMA924EU-12MM MMMA924EU-19MM
Adhesive transfer tapes, ATG 924, Scotch®
Tape
Apply Scotch® ATG adhesive transfer tape 924 for various substrate or material combinations. This transfer film tape is an acrylic, reverse wound, pressure sensitive adhesive on a densified kraft paper liner. Use with Scotch® ATG adhesive applicators.
  • Provides a clean, permanent bond to lightweight materials
  • Get high adhesion, good solvent resistance, and excellent ultraviolet (UV) resistance
  • Bonds on contact, and works on a variety of substrate or material combinations
  • Leaves no mess, no residue, and no odor
  • Length: 33 m
  • Transfer tape back material
This transfer film tape is a great general-purpose adhesive tape, offering high adhesion, good solvent resistance, and excellent ultraviolet resistance. It bonds on contact when delivered through the ATG system. A controlled application of the adhesive is delivered while the liner rewinds into the ATG applicator. The ATG tape system is a cost effective replacement to rivets, staples, screws, bolts, or liquid adhesives, and is safer than hot melt adhesives. It’s versatile, convenient, quick, and easy to use.