Industrial masking tapes, 401E
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4017250 401E-18MM 401E-24MM 401E-36MM 401E-48MM
MMMA4017250EA 2330 DKK
MMMA4017250 MMMA401E-18MM MMMA401E-24MM MMMA401E-36MM MMMA401E-48MM
Industrial masking tapes, 401E
Tape
401E masking tape is a smooth crepe paper tape with rubber adhesive. It offers high tensile strength, with a thickness of 0,170 mm.
  • Ideal in painting baking processes
  • May be exposed to temperatures up to 140 °C
  • Will not tear
  • Resistant to solvents and moisture, high temperatures
  • Designed for indoor use
  • Tensile strength: 100 [Newton/25 mm]
  • Crepe paper back material
Suitable for all oven-drying systems, infrared, or heat lamps.