General-purpose low noise box sealing tape 309, Scotch®
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
309-BR 309-CL
817-0439EA 43.9 DKK
817-0439 817-0440
General-purpose low noise box sealing tape 309, Scotch®
Tape
Scotch® tape is a general purpose box sealing tape with a polypropylene film backing and acrylic adhesive. The polypropylene backing is conformable, providing edge tear and split resistance.
  • General purpose packaging tape suitable for lightweight and price-sensitive applications
  • Low noise dispensing
  • Resists UV rays without yellowing for archival applications
  • High edge tear and split resistance
This is a 50 µm, general purpose polypropylene film tape with pressure-sensitive acrylic adhesive that offers reliability in lightweight and price-sensitiveMore Product Information