Filament tape 8953, Tartan™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8953-18MM 8953-25MM 8953-50MM
MMMA8953-18MMEA 33.4 DKK
MMMA8953-18MM MMMA8953-25MM MMMA8953-50MM
Filament tape 8953, Tartan™
Tape
Long life filament adhesive tape. This tape may be used for many applications such as securing a pallet package, assembling products, or closing and reinforcing packaging cartons. Once glued, the tape retains its adhesive strength under all temperatures it may face during transport and storage.
  • General use in cardboard reinforcement, light wrapping, or maintenance
  • Polypropylene film, fibreglass reinforced
  • Good initial tack to most fibreboard, plastics, and metal surfaces
  • Transparency provides clear visibility of box graphics, printing, and illustrations
  • Tape resists nicks, abrasions, moisture, and scuffing
The main application is the wrapping of light products.