General-purpose masking tape 201E
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
201E-18MM 201E-24MM 201E-30MM 201E-36MM 201E-48MM
MMMA201E-18MMEA 24.6 DKK
MMMA201E-18MM MMMA201E-24MM MMMA201E-30MM MMMA201E-36MM MMMA201E-48MM
General-purpose masking tape 201E
Tape
This is a premium-quality masking tape used for short-term paint masking, fixing, bundling, holding, sealing, and other applications up to a temperature of 80 °C for one hour.
  • Good instant adhesion and holding power that resists lifting or curling
  • Operating temperature range up to 80 °C for one hour
  • Reduces paint build-up and provides good paint lines, helps resist paint bleed through
  • Designed for indoor use
This tape is designed for general use in industrial applications with a slight need for temperature and water resistance, such as short-term paint masking and sealing applications. It provides a good instant adhesion and holding power that resists lifting or curling. Crepe paper backing offers conformability that will maintain integrity when formed around corners. Good solvent and moisture resistance helps to resist paint bleed through and provides good paint lines. It offers clean removability in one piece with no residue from most surfaces.