Dual Lock™ SJ354D Reclosable Fastener Twin Pack
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SJ354B SJ354D
293-0005EA 416 DKK
293-0005 293-0014
Dual Lock™ SJ354D Reclosable Fastener Twin Pack
Klæbemiddel
This reclosable fastener has a rubber based adhesive on the back of a black, polypropylene reclosable fastener. It's suitable for bonding many plastics, powder coated paints, and other low surface energy materials.
  • Snap-in-place installation replaces screws, nails, snaps, and rivets
  • Type 250 mates to Type 170, 250, or 400
  • Closes with an audible snap
  • Opens and closes 1000 times before losing 50% of original tensile strength
  • Pressure-sensitive adhesive provides excellent initial tack to most surfaces
  • Helps reduce vibration
Designed for use with many plastics (acrylic, polycarbonate, and AS), powder coated paints, and low surface energy materials (polypropylene and polyethylene). It's a type 250 stem density polyolefin fastener that is backed with a rubber based, pressure sensitive adhesive. The reclosable fastener has 40 mushroom shaped stems/cm² with a 5,8 mm engaged thickness. This product has low temperature performance, with a temperature resistance up to 50 °C long-term (70 °C short-term), and is therefore intended for indoor use only.