Non-phthalate PVC vacuum tubing, Nalgene™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8702-0055 8702-0065 8702-1065 8702-0145 8702-1145 8702-0210 8702-1210
NALG8702-0055EA 2010 DKK
NALG8702-0055 NALG8702-0065 NALG8702-1065 NALG8702-0145 NALG8702-1145 NALG8702-0210 NALG8702-1210
Non-phthalate PVC vacuum tubing, Nalgene™
Slanger Vakuum-/Trykslanger
Nalgene™ lab-grade, autoclavable clear tubing sets the standard for purity and quality, made from highest-grade resins, free from ortho-phthalate plasticisers.
  • Compare to Tygon® E-3603 tubing
  • Marked with Int.Ø and regulatory compliance as well as at 305 mm intervals for easy identification and measurement
  • Durometer hardness is shore A, 60
  • Thick-walled PVC tubing withstands a full vacuum of 760 mm Hg at room temperature
Nalgene™ Non-phthalate PVC vacuum tubing is manufactured from the highest-grade resin materials and performs like classic Nalgene™ 180 tubing but is free from orthophthalate DEHP plasticisers. Resists hardening and discolouration from a wide range of laboratory chemicals (demonstrates dimensional stability) and does not impart odour or taste.
Certificeringer: Meets food additives amendment of the federal food, drug, and cosmetic act. Meets the requirements of the U.S. pharmacopoeia class VI biological tests for plastics, USP <88>. Complies with FDA CFR21 for food packaging, USP VI, cytotoxicity, REACH, latex-free, RoHS, and California proposition 65 standards.
Advarsel: Not intended for medical use.