Chlorobenzene : Acetic Acid solution (2:1) (v/v), Burdick & Jackson™
Leverandør: Honeywell Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
66163-200L
66163-2.5L
HONC66163-200LEA
45110
DKK
HONC66163-200L
HONC66163-2.5L
Chlorobenzene : Acetic Acid solution (2:1) (v/v), Burdick & Jackson™
Chlorobenzene : Acetic Acid
Kogepunkt:
118 °C Massefylde: 1,079 g/cm3 (20 °C) Flammepunkt: 27 °C Opbevaringstemperatur: Stue temperatur |
UN nummer:
3286 ADR: 3,II |
Specifikationer Testresultater
Assay of acetic acid | 31.5 - 33.5 % |
Density (D 20/4) | 1.070 - 1.090 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...