Laboratory coats, Tyvek® 500
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYPL39SWHNP/L TYPL39SWHNP/M TYPL39SWHNP/S TYPL39SWHNP/XL TYPL39SWHNP/XX
DUPOTYPL39SWHNP/LEA 3630 DKK
DUPOTYPL39SWHNP/L DUPOTYPL39SWHNP/M DUPOTYPL39SWHNP/S DUPOTYPL39SWHNP/XL DUPOTYPL39SWHNP/XX
Laboratory coats, Tyvek® 500
Engangsbeklædning
DuPont™ Tyvek® 500 Labcoat with zipper, model PL309NP, offers enhanced protection for individuals exposed to hazardous substances.
  • Stitched internal seams
  • Mandarin collar
  • Zipper front closure
  • Elasticated cuffs
  • Without pockets
Tyvek® 500 laboratory coats are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability, and comfort. Tyvek® material is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 micron), is ultra-low-linting, and antistatically treated. Silicon non-added.
Garment applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting, and general clean-up.
Basis weight is 41.5 g/m2, per test method DIN EN ISO 536.
Certificeringer: Certified according to Regulation (EU) 2016/425. Partial body chemical protective clothing, Category III, Type PB [6-B]. EN 14126. Antistatic treatment (EN 1149-1) - on both sides.