VWR®, Wire baskets
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
215-4200EA 617 DKK
215-4200 215-4201 215-4202 215-4203 215-4204 215-4205 215-4206 215-4207
VWR®, Wire baskets
Kurve Kurve, tråd
These wire baskets are made from rust-proof, electrolytically polished stainless steel.
  • Very good chemical and temperature resistance
  • Spot-welded mesh 8×8×1 mm
  • Reinforced edges
Can be used for transport, storage or sterilisation of laboratory utensils.