General purpose gloves, nitrile, Showa S-TEX 376R
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
STEX376-9/XL STEX376-10/XXL STEX376-6/S STEX376-7/M STEX376-8/L
112-6011EA 1990 DKK
112-6011 112-6007 112-6008 112-6009 112-6010
General purpose gloves, nitrile, Showa S-TEX 376R
Handsker
Showa S-TEX 376 durable double-dipped nitrile coated glove that provides resistance to oil whilst protecting from cuts and lacerations. Ideal for the secure handling of oily components and materials that present a high degree of mechanical risk. Manufactured with SHOWA's patented Hagane Coil® technology.
  • ¾ coated nitrile with an additional coating of foam nitrile on the palm, fingertips and knuckles
  • Protects the hand from oils, hydrocarbons, grease and abrasion
  • Long lasting grip performance in wet and oily conditions
  • Anatomical hand shape design helps prevent hand fatigue
  • Seamless nylon/polyester knitted liner
  • High mechanical resistance to abrasion
Idea for use in aerospace, automotive, construction, oil and gas, utilities and waste management. Typical application includes assembly, handling oily/greasy components, maintenance, metal fabrication, mixing mortar and refuse collection.
Certificeringer: EN 388:2016 (4121X); Cat. II