General purpose gloves, Showa 381
Leverandør: Showa
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
381-6/S 381-7/M 381-8/L 381-9/XL 381-10/XXL
112-5826EA 761 DKK
112-5826 112-5827 112-5828 112-5829 112-5825
General purpose gloves, Showa 381
Handsker
SHOWA 381 is a general purpose, durable, lightweight, foam nitrile coated, with a breathable microfibre knit liner and extremely comfortable gloves. Ideal for assembly and precision handling tasks.
  • Microporous nitrile coating allows warm air and moisture from inside to escape
  • Excellent grip in light oily and dry environments
  • Microfibre yarn absorbs perspiration and moisture from the skin, resulting in drier and cooler hands
  • Excellent levels of tactility and sensitivity
  • Exceptional suppleness, comfort and fit
  • Abrasion level 4 for maximum abrasion resistance
Ideal for use in construction, maintenance, logistics, aerospace, automotive, engineering and manufacturing. Typical applications include: Assembly of dry and oily parts, general handling, handling of intricate components, light fabrication, mounting and dismounting of parts and tool setting.
Certificeringer: EN 388:2016 (4121X); Cat. II