VWR®, Single-Use Transfer Hose with SterilEnz™ Connections
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
734-3323EA 2850 DKK
734-3323 734-3324
VWR®, Single-Use Transfer Hose with SterilEnz™ Connections
Slanger Overførselsslange
Avantor's high pressure single-use transfer hose with Avantor SterilEnz™ connections maintain the durability of high pressure hose products while providing the benefits and flexibility of single-use assemblies.
  • Suitable for elevated temperatures and pressures up to 120 psi
  • 25,4 mm inner diameter for high flow rate applications
  • Gamma irradiated with SAL 10⁻⁶
  • Offered at 1,524 and 3,048 m lengths
  • Gasket affixed to TC face even when disconnected to prevent misalignment
Designed for applications where pressures would exceed normal biopharmaceutical processes.
Avantor single-use transfer hose with SterilEnz™ connections are pre-sterilised and gamma irradiated with SAL 10⁻⁶, then packed in a double-polybag as an added convenience for customers when working with the product. The inner diameter supports high flow rates, while the different lengths meet the requirements for many bioprocessing applications, with custom lengths available upon request.
Certificeringer: USP Class VI, ACDF, Sterile, Pressure rating up to 120 psi.