Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
FD001X34 ODNABC33 ODNABC34
516-9341EA 1200 DKK
516-9341 PALLODNABC33 PALLODNABC34
Centrifugal devices for RNA and DNA extraction, NAB Nanosep®
Filtre Centrifugefiltre
The Pall Nanosep device for nucleic acid binding (NAB) incorporates a dual layer silica-based quartz glass fiber media to allow for efficient binding of DNA and RNA, while providing smooth flow and rapid processing of samples.
  • Rapid processing of samples
  • Silica-based quartz glass fiber media that allows efficient binding of DNA and RNA
  • Constructed of low-binding polypropylene
  • Ultrasonically welded seals prevent by passor seal failure
  • Fits standard centrifuge rotors that accept 1,5 ml tubes
This media offers researchers the flexibility to purifyMore Product Information
Bestil nu
Specifikationer
Environmentally Preferable
Filtreringsareal 0.28 cm²
Max. RCF 14,000 ×g
Ø×H 45×23 mm
Prøvevolumen max. 500 µl
Temperatur 15 - 30 °C (59 - 86 °F)