Sealing films, Seal-R-film™
Leverandør: Brand
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
701620 701624 701626 701622
219-0000EA 411 DKK
219-0000 219-0002 219-0003 219-0001
Sealing films, Seal-R-film™
Forsegling Plastfolie
Seal-R-film™ sealing film seals beakers, flasks, and other vessels.
  • Protects against evaporation, leakage, and contamination
  • Easy sealing with 300% elasticity
  • Lot identification of each roll
The sealing film can be stretched to three times its original length. This means that Seal-R-film™ always fits, even with irregular shapes. For traceability, each roll has a lot identification. With the BRAND cutter for sealing film you can cut rolls with widths of 50 and 100 mm efficiently and economically and store them cleanly.
Specifications:
- Thickness: 0.14 +/- 0.02mm
- Toxicity: non-toxic
- Melting point: > 66°C (151°F)
- Boiling point: > 288°C (550°F)
- Continuous service temperature: –20 °C to 46 °C
- Elasticity (25°C): 300%
- Tear strength (25°C): 1.5 MPa
Gas permeability in 24h at room temperature and a pressure of 0.1 MPa:
- CO2 (carbon dioxide) < 1000 cm³/m²
- O2 (oxygen) < 350 cm³/m²
- Water vapor permeability < 0.8 g/m²
Chemical resistance:
- Hydrochloric acid (HCL) 36.5% - resistant
- Sulfuric acid (H2SO4) 98% - resistant
- Nitric acid (HNO3) 95% - resistant*
- Hydrogen peroxide (H202) 30% - resistant
- Sodium hydroxide (NaOH) 22% - resistant
- Ammonia (NH3) 28% - resistant
- Sodium chloride (NaCl) 20% - resistant
- Potassium permanganate (KMnO4) 5% - resistant*
- Iodine solution 0.1 mol/l - resistant*
- Methanol - resistant
- Ethanol - resistant
- Isopropanol - resistant
* Brown coloration