Tetrabutylammoniumhydroxid 20% in aqueous solution, GPR RECTAPUR®
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
85740.180EA 398 DKK
85740.180 85740.260
Tetrabutylammoniumhydroxid 20% in aqueous solution, GPR RECTAPUR®
Tetrabutylammonium hydroxide
Formel: C₁₆H₃₇NO
Molekylvægt: 259.48 g/mol
Smeltepunkt: 29.85
Massefylde: 0.968
MDL Number: MFCD00009425
CAS-nummer: 2052-49-5
UN nummer: 3267
ADR: 8 II